Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NGFR SIRNA Assays ›
Gene Symbol:
Ngfr
Gene Name:
nerve growth factor receptor
Gene Aliases:
LNGFR, RNNGFRR, Tnfrsf16, p75, p75NTR
Chromosome Location:
Chr.10: 83389835 - 83408061 on Build Rnor_6.0
UniGene ID:
Rn.10980
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
47950
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Ngfr
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_012610.2
NP_036742.2
2
203
GenBank mRNA
CB725789.1
-
2
211
X05137.1
-
2
203
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_033217, AF105292, BB630380, BC038365, AK037248.
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
intracellular protein transport
apoptotic process
signal transduction
axon guidance
central nervous system development
response to wounding
regulation of gene expression
positive regulation of cell death
negative regulation of neuron projection development
programmed cell death
detection of temperature stimulus
sensory perception of pain
nerve development
hair follicle morphogenesis
positive regulation of myelination
positive regulation of synaptic transmission, cholinergic
response to lipopolysaccharide
circadian regulation of gene expression
cellular response to oxidative stress
positive regulation of Rho protein signal transduction
skeletal muscle cell differentiation
negative regulation of fibroblast growth factor receptor signaling pathway
positive regulation of odontogenesis of dentin-containing tooth
glucose homeostasis
positive regulation of apoptotic process
positive regulation of MAPK cascade
negative regulation of neuron apoptotic process
skin development
positive regulation of fibroblast proliferation
negative regulation of hair follicle development
positive regulation of protein kinase B signaling
negative regulation of mitochondrial depolarization
positive regulation of synaptic transmission, glutamatergic
negative regulation of cell death
positive regulation of neuron death
regulation of reactive oxygen species metabolic process
positive regulation of apoptotic signaling pathway
regulation of glucose import in response to insulin stimulus
beta-amyloid binding
death receptor activity
neurotrophin TRKA receptor binding
protein binding
Rab GTPase binding
ubiquitin protein ligase binding
neurotrophin binding
nerve growth factor binding
preprotein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-mw5n9:80/100.66.77.197:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline