Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012623.2 | NP_036755.2 | 3 | 115 | ||
XM_008762687.1 | XP_008760909.1 | 2 | 253 | |||
XM_008762688.1 | XP_008760910.1 | 2 | 252 | |||
XM_008762691.1 | XP_008760913.1 | 2 | 247 | |||
GenBank mRNA | AY082609.1 | - | 3 | 115 | ||
BC107560.1 | - | 3 | 215 | |||
M81855.1 | - | 3 | 182 |