Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SPARC SIRNA Assays ›
          Gene Symbol:
          Sparc
          Gene Name:
          secreted protein, acidic, cysteine-rich (osteonectin)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 40742390 - 40764222 on Build Rnor_6.0
          UniGene ID:
          Rn.234316
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 47991

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Sparc
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012656.1 NP_036788.1 5 200
          XM_008767663.1 XP_008765885.1 4 278
          GenBank mRNA BC061777.1 - 4 215 5621666
          D28875.1 - 4 217
          FQ212364.1 - 4 269
          FQ212499.1 - 4 234
          FQ213815.1 - 4 234
          FQ213945.1 - 4 234
          FQ214222.1 - 4 232
          FQ214492.1 - 4 234
          FQ214649.1 - 4 234
          FQ214673.1 - 4 234
          FQ215195.1 - 4 233
          FQ215581.1 - 4 235
          FQ215622.1 - 4 236
          FQ215818.1 - 4 245
          FQ216823.1 - 4 236
          FQ217546.1 - 4 235
          FQ219866.1 - 4 216
          FQ219932.1 - 5 216
          FQ219983.1 - 4 234
          FQ220003.1 - 4 234
          FQ220043.1 - 4 232
          FQ220046.1 - 4 234
          FQ220186.1 - 4 232
          FQ220279.1 - 2 58
          FQ220287.1 - 4 236
          FQ220443.1 - 4 216
          FQ220507.1 - 4 242
          FQ220547.1 - 4 234
          FQ220572.1 - 4 234
          FQ220644.1 - 4 234
          FQ220827.1 - 4 234
          FQ220880.1 - 4 234
          FQ220901.1 - 4 234
          FQ222224.1 - 4 234
          FQ223025.1 - 4 234
          FQ225033.1 - 4 236
          FQ225092.1 - 4 230
          FQ225200.1 - 4 234
          FQ225220.1 - 4 213
          FQ225486.1 - 4 216
          FQ225609.1 - 4 234
          FQ225618.1 - 4 232
          FQ225678.1 - 4 234
          FQ225688.1 - 4 234
          FQ225694.1 - 4 236
          FQ225722.1 - 4 232
          FQ225884.1 - 4 232
          FQ225913.1 - 4 234
          FQ225914.1 - 4 236
          FQ225965.1 - 4 234
          FQ226299.1 - 4 216
          FQ226354.1 - 4 234
          FQ226360.1 - 4 234
          FQ226584.1 - 4 234
          FQ226599.1 - 4 235
          FQ226651.1 - 4 257
          FQ226673.1 - 4 215
          FQ226731.1 - 4 234
          FQ226746.1 - 4 232
          FQ227279.1 - 4 234
          FQ227628.1 - 4 242
          FQ227644.1 - 4 234
          FQ227708.1 - 4 234
          FQ227725.1 - 4 234
          FQ227727.1 - 4 213
          FQ227749.1 - 4 234
          FQ228032.1 - 4 214
          FQ228597.1 - 4 216
          FQ229172.1 - 4 234
          FQ229199.1 - 4 234
          FQ229230.1 - 4 234
          FQ229283.1 - 4 216
          FQ229315.1 - 4 232
          FQ229316.1 - 4 234
          FQ230029.1 - 4 234
          FQ230086.1 - 4 232
          FQ230210.1 - 4 235
          FQ230381.1 - 4 234
          FQ230595.1 - 4 216
          FQ230619.1 - 4 234
          FQ230667.1 - 4 234
          FQ230705.1 - 4 232
          FQ230778.1 - 4 234
          FQ230864.1 - 4 234
          FQ230993.1 - 4 234
          FQ231067.1 - 4 234
          FQ231166.1 - 4 233
          FQ231444.1 - 4 233
          FQ231523.1 - 4 234
          FQ231543.1 - 4 234
          FQ231632.1 - 4 234
          FQ231753.1 - 4 231
          FQ231815.1 - 4 216
          FQ231918.1 - 4 216
          FQ231975.1 - 4 216
          FQ232042.1 - 4 233
          FQ232191.1 - 4 234
          FQ232201.1 - 4 234
          FQ232223.1 - 4 234
          FQ232271.1 - 4 232
          FQ232364.1 - 4 236
          FQ232445.1 - 4 232
          FQ232464.1 - 4 234
          FQ232710.1 - 4 232
          FQ232838.1 - 4 234
          FQ232847.1 - 4 234
          FQ232862.1 - 4 232
          FQ232869.1 - 4 234
          FQ232965.1 - 4 234
          FQ232977.1 - 4 234
          FQ232986.1 - 4 234
          FQ233178.1 - 4 234
          FQ233283.1 - 4 213
          FQ233669.1 - 4 233
          FQ233698.1 - 4 216
          FQ233777.1 - 4 232
          FQ233810.1 - 4 236
          FQ233845.1 - 4 234
          FQ233850.1 - 4 235
          FQ233926.1 - 4 234
          FQ234037.1 - 4 234
          FQ234069.1 - 4 234
          FQ234108.1 - 4 234
          FQ234199.1 - 4 234
          FQ234410.1 - 4 216
          FQ234523.1 - 4 221
          FQ234589.1 - 4 232
          FQ234657.1 - 4 216
          FQ234685.1 - 4 234
          FQ234766.1 - 4 231
          FQ235008.1 - 4 236
          FQ235038.1 - 4 234
          FQ235049.1 - 4 216
          Y13714.1 - 5 200

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          extracellular matrix glycoprotein

          Gene Ontology Categories:

          Function(s) Process(es)

          ossification
          negative regulation of endothelial cell proliferation
          signal transduction
          heart development
          response to gravity
          response to lead ion
          positive regulation of endothelial cell migration
          negative regulation of angiogenesis
          regulation of cell morphogenesis
          lung development
          response to lipopolysaccharide
          response to L-ascorbic acid
          response to cytokine
          wound healing
          regulation of cell proliferation
          response to peptide hormone
          response to ethanol
          response to cadmium ion
          inner ear development
          response to glucocorticoid
          response to cAMP
          response to calcium ion
          bone development
          cellular response to growth factor stimulus
          calcium ion binding
          collagen binding
          extracellular matrix binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-d2m8d:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0