Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MYLPF SIRNA Assays ›
          Gene Symbol:
          Mylpf
          Gene Name:
          myosin light chain, phosphorylatable, fast skeletal muscle
          Gene Aliases:
          MLC2, Myl2, Myolc1
          Chromosome Location:
          Chr.1: 198655835 - 198658590 on Build Rnor_6.0
          UniGene ID:
          Rn.6534
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 48128

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Mylpf
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012605.2 NP_036737.1 2, 3 145
          GenBank mRNA CB770854.1 - 2, 3 155
          FQ215085.1 - 2, 3 178
          FQ216891.1 - 2, 3 161
          FQ216934.1 - 2, 3 161
          FQ217090.1 - 2, 3 163
          FQ217098.1 - 2, 3 161
          FQ217155.1 - 2, 3 161
          FQ217219.1 - 2, 3 161
          FQ217259.1 - 2, 3 161
          FQ217349.1 - 1, 2 85
          FQ217365.1 - 2, 3 160
          FQ217368.1 - 2, 3 177
          FQ217533.1 - 2, 3 189
          FQ217703.1 - 2, 3 161
          FQ217740.1 - 2, 3 177
          FQ217881.1 - 2, 3 158
          FQ217944.1 - 2, 3 161
          FQ218017.1 - 2, 3 161
          FQ223762.1 - 2, 3 142
          FQ223868.1 - 2, 3 161
          FQ223911.1 - 2, 3 177
          FQ223944.1 - 1, 2 27
          FQ223966.1 - 2, 3 161
          FQ223985.1 - 2, 3 161
          FQ224231.1 - 2, 3 161
          FQ224253.1 - 2, 3 160
          FQ224391.1 - 2, 3 159
          FQ224404.1 - 2, 3 159
          FQ224511.1 - 2, 3 161
          FQ224525.1 - 2, 3 180
          FQ224720.1 - 2, 3 161
          FQ224781.1 - 2, 3 177

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          muscle contraction
          immune response
          skeletal muscle tissue development
          calcium ion binding
          structural constituent of muscle

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-9c8cf:80/100.66.75.150:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0