Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GCG SIRNA Assays ›
Gene Symbol:
Gcg
Gene Name:
glucagon
Gene Aliases:
GLP-1
Chromosome Location:
Chr.3: 48442635 - 48451650 on Build Rnor_6.0
UniGene ID:
Rn.54383
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
48205
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
286.00
Your Price
Online offer:
277.65
277.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Gcg
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_012707.2
NP_036839.2
3, 4
320
GenBank mRNA
BP479145.1
-
3, 4
320
BP480555.1
-
3, 4
295
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_008100, BC012975, AF276754, AK007911, Z46845, and others.
Gene Ontology Categories:
Function(s)
Process(es)
regulation of carbohydrate metabolic process
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
protein kinase A signaling
positive regulation of peptidyl-threonine phosphorylation
regulation of lipid metabolic process
protein metabolic process
positive regulation of cAMP biosynthetic process
positive regulation of protein binding
negative regulation of appetite
positive regulation of peptidyl-serine phosphorylation
positive regulation of insulin secretion involved in cellular response to glucose stimulus
positive regulation of gluconeogenesis by positive regulation of transcription from RNA polymerase II promoter
response to starvation
negative regulation of apoptotic process
positive regulation of protein kinase activity
regulation of insulin secretion
positive regulation of histone H3-K4 methylation
positive regulation of ERK1 and ERK2 cascade
positive regulation of calcium ion import
negative regulation of execution phase of apoptosis
negative regulation of intrinsic apoptotic signaling pathway
hormone activity
glucagon receptor binding
identical protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline