Search
Search

Mfge8| Gene Symbol: Mfge8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040186.2 | NP_001035276.1 | 2 | 249 | ||
| NM_012811.3 | NP_036943.1 | 2 | 249 | |||
| GenBank mRNA | BC085754.1 | - | 2 | 202 | 7193796 | |
| CO404762.1 | - | 2 | 251 | |||
| D84068.1 | - | 2 | 209 | |||
| DQ455823.1 | - | 2 | 202 | |||
| FQ134652.1 | - | 2 | 249 | |||