Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CYCS SIRNA Assays ›
          Gene Symbol:
          Cycs LOC100363502
          Gene Name:
          cytochrome c, somatic
          cytochrome c, somatic-like
          Gene Aliases:
          CYCSA
          -
          Chromosome Location:
          Chr.4: 80331226 - 80333326 on Build Rnor_6.0
          Chromosome Location:
          Chr.18: 62475572 - 62476771 on Build Rnor_6.0
          UniGene ID:
          Rn.161841
          -
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 48488

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Cycs
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012839.2 NP_036971.1 2 153
          GenBank mRNA BC081849.1 - 2 153 7192255
          BC089051.1 - 2 164 7305562
          FQ211631.1 - 2 145
          FQ212556.1 - 2 147
          FQ212860.1 - 2 205
          FQ213464.1 - 2 145
          FQ214646.1 - 2 176
          FQ214733.1 - 2 145
          FQ214738.1 - 2 143
          FQ214838.1 - 2 145
          FQ214855.1 - 2 176
          FQ214908.1 - 2 145
          FQ215052.1 - 2 145
          FQ215109.1 - 2 145
          FQ215466.1 - 2 145
          FQ215858.1 - 2 188
          FQ216163.1 - 2 144
          FQ216217.1 - 2 143
          FQ216281.1 - 2 203
          FQ216426.1 - 2 145
          FQ216512.1 - 2 145
          FQ216560.1 - 2 145
          FQ216674.1 - 2 144
          FQ216679.1 - 2 145
          FQ217283.1 - 2 145
          FQ217634.1 - 2 145
          FQ218428.1 - 2 145
          FQ218668.1 - 2 146
          FQ220222.1 - 2 176
          FQ220855.1 - 2 188
          FQ223761.1 - 2 145
          FQ224710.1 - 2 145
          FQ224914.1 - 2 188
          FQ224919.1 - 2 145
          M20622.1 - 1 78
          Gene Symbol: LOC100363502
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_003751799.3 XP_003751847.1 1 149

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          response to ischemia
          mitochondrial electron transport, ubiquinol to cytochrome c
          mitochondrial electron transport, cytochrome c to oxygen
          apoptotic DNA fragmentation
          apoptotic process
          response to oxidative stress
          activation of cysteine-type endopeptidase activity involved in apoptotic process by cytochrome c
          negative regulation of hydrogen peroxide biosynthetic process
          glial cell apoptotic process
          hydrogen peroxide metabolic process
          positive regulation of cellular respiration
          iron ion binding
          lipid binding
          electron carrier activity
          heme binding
          electron transporter, transferring electrons from CoQH2-cytochrome c reductase complex and cytochrome c oxidase complex activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0