Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Sftpd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012878.2 | NP_037010.1 | 2 | 285 | ||
XM_008771065.1 | XP_008769287.1 | 2 | 288 | |||
GenBank mRNA | BC070507.1 | - | 3 | 294 | 7098194 | |
M81231.1 | - | 3 | 292 |