Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HMGCR SIRNA Assays ›
Gene Symbol:
Hmgcr
Gene Name:
3-hydroxy-3-methylglutaryl-CoA reductase
Gene Aliases:
3H3M
Chromosome Location:
Chr.2: 27480224 - 27500654 on Build Rnor_6.0
UniGene ID:
Rn.9437
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
49086
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Hmgcr
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_013134.2
NP_037266.2
4, 5
416
XM_006231826.2
XP_006231888.1
4, 5
824
GenBank mRNA
BC064654.1
-
4, 5
416
5600023
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_008255, BC085083, AK159899.
Panther Classification:
Molecular Function -
oxidoreductase
reductase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
Process(es)
cholesterol biosynthetic process
ubiquinone metabolic process
aging
response to nutrient
positive regulation of cell proliferation
isoprenoid biosynthetic process
visual learning
farnesyl diphosphate biosynthetic process, mevalonate pathway
negative regulation of striated muscle cell apoptotic process
positive regulation of cardiac muscle cell apoptotic process
coenzyme A metabolic process
positive regulation of stress-activated MAPK cascade
negative regulation of apoptotic process
negative regulation of MAP kinase activity
myoblast differentiation
response to ethanol
negative regulation of vasodilation
positive regulation of skeletal muscle tissue development
positive regulation of smooth muscle cell proliferation
protein tetramerization
oxidation-reduction process
negative regulation of wound healing
negative regulation of insulin secretion involved in cellular response to glucose stimulus
positive regulation of ERK1 and ERK2 cascade
hydroxymethylglutaryl-CoA reductase (NADPH) activity
hydroxymethylglutaryl-CoA reductase activity
protein homodimerization activity
coenzyme binding
protein phosphatase 2A binding
NADPH binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0