Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CNTF SIRNA Assays ›
Gene Symbol:
Cntf
Gene Name:
ciliary neurotrophic factor
Gene Aliases:
-
Chromosome Location:
Chr.1: 229599009 - 229601032 on Build Rnor_6.0
UniGene ID:
Rn.6067
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
49275
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Cntf
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_013166.1
NP_037298.1
1
159
GenBank mRNA
FQ216326.1
-
1
122
X17457.1
-
1
159
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Human: NM_000614, BC074963, DB241497, AB057443, BC074964, and others.
This siRNA also targets the following transcripts in Mouse: NM_170786, BC027539, W08735, U05342, AK041204.
Gene Ontology Categories:
Function(s)
Process(es)
JAK-STAT cascade
nervous system development
positive regulation of cell proliferation
response to light intensity
positive regulation of phosphatidylinositol 3-kinase signaling
positive regulation of cell growth
growth
positive regulation of tyrosine phosphorylation of Stat3 protein
positive regulation of MAPK cascade
negative regulation of neuron apoptotic process
positive regulation of JAK-STAT cascade
negative regulation of photoreceptor cell differentiation
regulation of retinal cell programmed cell death
positive regulation of hormone secretion
astrocyte activation
muscle organ morphogenesis
neuron development
positive regulation of axon regeneration
protein heterooligomerization
response to electrical stimulus
regulation of resting membrane potential
membrane hyperpolarization
ciliary neurotrophic factor-mediated signaling pathway
negative regulation of neuron death
cytokine activity
ciliary neurotrophic factor receptor binding
interleukin-6 receptor binding
growth factor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System