Search
Search

Pdlim4| Gene Symbol: Pdlim4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017062.2 | NP_058758.1 | 2 | 269 | ||
| XM_006246230.2 | XP_006246292.1 | 2 | 614 | |||
| XM_006246231.2 | XP_006246293.1 | 2 | 327 | |||
| GenBank mRNA | X76454.1 | - | 2 | 270 | ||