Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rltpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033320.2 | NP_001028492.1 | 13, 14 | 1996 | ||
GenBank mRNA | AK031012.1 | - | 13, 14 | 1995 | ||
AK051288.1 | - | 1, 2 | 634 | |||
BC115830.1 | - | 4, 5 | 1702 | |||
HF678090.1 | - | 34, 35 | 3627 |