Search Thermo Fisher Scientific
Gene Symbol: Card6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163138.1 | NP_001156610.1 | 4 | 4208 | ||
GenBank mRNA | AK052796.1 | - | 3 | 4098 | ||
BC038655.1 | - | 1 | 429 |