Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Dgki | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081206.1 | NP_001074675.1 | 22 | 2256 | ||
GenBank mRNA | AK081360.1 | - | 22 | 2259 | ||
AK086550.1 | - | 8 | 785 |