Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PRDM10 SIRNA Assays ›
          Gene Symbol:
          Prdm10
          Gene Name:
          PR domain containing 10
          Gene Aliases:
          Gm1112, tristanin
          Chromosome Location:
          Chr.9: 31278780 - 31381731 on Build GRCm38
          UniGene ID:
          Mm.335039
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 504278

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Prdm10
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001080817.1 NP_001074286.1 16 2622

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_199438, NM_020228, NM_199439, NM_199437, AB033057, and others.

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          biological_process
          methylation
          rRNA (adenine-N6,N6-)-dimethyltransferase activity
          nucleic acid binding
          DNA binding
          protein C-terminal carboxyl O-methyltransferase activity
          tRNA (guanine-N2-)-methyltransferase activity
          methyltransferase activity
          C-methyltransferase activity
          N-methyltransferase activity
          O-methyltransferase activity
          S-methyltransferase activity
          RNA methyltransferase activity
          mRNA methyltransferase activity
          tRNA methyltransferase activity
          protein methyltransferase activity
          2-polyprenyl-6-methoxy-1,4-benzoquinone methyltransferase activity
          rRNA methyltransferase activity
          rRNA (uridine-2'-O-)-methyltransferase activity
          rRNA (adenine-N6-)-methyltransferase activity
          DNA-methyltransferase activity
          tRNA (guanine-N1-)-methyltransferase activity
          rRNA (cytosine-C5-)-methyltransferase activity
          selenocysteine methyltransferase activity
          arginine N-methyltransferase activity
          protein-arginine N-methyltransferase activity
          lysine N-methyltransferase activity
          protein-lysine N-methyltransferase activity
          tRNA (uracil) methyltransferase activity
          tRNA (guanine) methyltransferase activity
          tRNA (adenine) methyltransferase activity
          tRNA (cytosine) methyltransferase activity
          tRNA (cytosine-5-)-methyltransferase activity
          tRNA (adenine-N1-)-methyltransferase activity
          rRNA (adenine) methyltransferase activity
          rRNA (cytosine) methyltransferase activity
          rRNA (guanine) methyltransferase activity
          rRNA (uridine) methyltransferase activity
          transferase activity
          protein C-terminal leucine carboxyl O-methyltransferase activity
          1-phenanthrol methyltransferase activity
          protein-arginine N5-methyltransferase activity
          methylarsonite methyltransferase activity
          dimethylarsinite methyltransferase activity
          4,5-dihydroxybenzo(a)pyrene methyltransferase activity
          1-hydroxypyrene methyltransferase activity
          1-hydroxy-6-methoxypyrene methyltransferase activity
          demethylmenaquinone methyltransferase activity
          cobalt-precorrin-6B C5-methyltransferase activity
          cobalt-precorrin-7 C15-methyltransferase activity
          cobalt-precorrin-5B C1-methyltransferase activity
          cobalt-precorrin-3 C17-methyltransferase activity
          dimethylamine methyltransferase activity
          hydroxyneurosporene-O-methyltransferase activity
          tRNA (adenine-57, 58-N(1)-) methyltransferase activity
          methylamine-specific methylcobalamin:coenzyme M methyltransferase activity
          trimethylamine methyltransferase activity
          methanol-specific methylcobalamin:coenzyme M methyltransferase activity
          monomethylamine methyltransferase activity
          metal ion binding
          P-methyltransferase activity
          Se-methyltransferase activity
          2-phytyl-1,4-naphthoquinone methyltransferase activity
          tRNA (uracil-2'-O-)-methyltransferase activity
          tRNA (cytosine-2'-O-)-methyltransferase activity
          phosphomethylethanolamine N-methyltransferase activity
          tRNA (cytosine-3-)-methyltransferase activity
          rRNA (cytosine-2'-O-)-methyltransferase activity
          rRNA (cytosine-N4-)-methyltransferase activity
          trihydroxyferuloyl spermidine O-methyltransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0