Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TLR4 SIRNA Assays ›
Gene Symbol:
Tlr4
Gene Name:
toll-like receptor 4
Gene Aliases:
-
Chromosome Location:
Chr.5: 82587424 - 82601056 on Build Rnor_6.0
UniGene ID:
Rn.14534
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
50434
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Tlr4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_019178.1
NP_062051.1
1
312
GenBank mRNA
AF057025.1
-
1
334
EU929065.1
-
1
37
GQ175880.1
-
1
312
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_021297, JX878358, AF110133, JX878359, AF185285, and others.
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
response to hypoxia
microglial cell activation
activation of innate immune response
toll-like receptor signaling pathway
B cell proliferation involved in immune response
nitric oxide production involved in inflammatory response
MyD88-dependent toll-like receptor signaling pathway
innate immune response-activating signal transduction
skeletal muscle contraction
leukotriene metabolic process
inflammatory response
response to oxidative stress
I-kappaB phosphorylation
response to bacterium
response to toxic substance
positive regulation of platelet activation
response to activity
immunoglobulin mediated immune response
positive regulation of B cell proliferation
lipopolysaccharide-mediated signaling pathway
response to lipopolysaccharide
detection of lipopolysaccharide
response to retinoic acid
response to progesterone
interferon-gamma production
negative regulation of interferon-gamma production
negative regulation of interleukin-17 production
negative regulation of interleukin-23 production
negative regulation of interleukin-6 production
negative regulation of tumor necrosis factor production
positive regulation of chemokine production
positive regulation of interferon-alpha production
positive regulation of interferon-beta production
positive regulation of interferon-gamma production
positive regulation of interleukin-1 production
positive regulation of interleukin-10 production
positive regulation of interleukin-12 production
positive regulation of interleukin-6 production
positive regulation of interleukin-8 production
positive regulation of tumor necrosis factor production
response to insulin
positive regulation of stress-activated MAPK cascade
toll-like receptor 4 signaling pathway
macrophage activation
positive regulation of NF-kappaB import into nucleus
positive regulation of tumor necrosis factor biosynthetic process
positive regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
response to morphine
positive regulation of DNA binding
positive regulation of interleukin-12 biosynthetic process
innate immune response
positive regulation of MHC class II biosynthetic process
positive regulation of interferon-beta biosynthetic process
positive regulation of interleukin-8 biosynthetic process
positive regulation of nitric oxide biosynthetic process
response to ethanol
positive regulation of transcription from RNA polymerase II promoter
positive regulation of JNK cascade
positive regulation of lymphocyte proliferation
regulation of cytokine secretion
regulation of inflammatory response
positive regulation of inflammatory response
positive regulation of peptidyl-tyrosine phosphorylation
defense response to Gram-negative bacterium
positive regulation of sequence-specific DNA binding transcription factor activity
positive regulation of NF-kappaB transcription factor activity
response to glucocorticoid
positive regulation of nitric-oxide synthase biosynthetic process
regulation of sensory perception of pain
intestinal epithelial structure maintenance
positive regulation of macrophage cytokine production
negative regulation of ERK1 and ERK2 cascade
positive regulation of ERK1 and ERK2 cascade
positive regulation of nucleotide-binding oligomerization domain containing 1 signaling pathway
positive regulation of nucleotide-binding oligomerization domain containing 2 signaling pathway
response to fatty acid
cellular response to lipopolysaccharide
cellular response to lipoteichoic acid
cellular response to mechanical stimulus
cellular response to retinoic acid
response to gold nanoparticle
lipopolysaccharide binding
lipopolysaccharide receptor activity
receptor activity
transmembrane signaling receptor activity
protein binding
phosphatidylinositol 3-kinase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline