Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Lipt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037918.3 | NP_001033007.2 | 2 | 1046 | ||
GenBank mRNA | AK048005.1 | - | 1047 | |||
BC019519.1 | - | 2 | 1035 | 4038179 |