Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Trim71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042503.2 | NP_001035968.1 | 4 | 4736 | ||
GenBank mRNA | AK011901.1 | - | 1 | 870 | ||
DQ005956.1 | - | 4 | 4736 |