Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Krtdap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033131.3 | NP_001028303.1 | 4, 5 | 245 | ||
GenBank mRNA | AK131603.1 | - | 4, 5 | 246 | ||
AK131606.1 | - | 4, 5 | 246 | |||
BC132510.1 | - | 4, 5 | 245 | 40130787 | ||
BC132512.1 | - | 4, 5 | 245 | 40130789 | ||
BC145437.1 | - | 3, 4 | 203 | 9053995 | ||
BC145438.1 | - | 4, 5 | 245 | 9053996 |