Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IL18 SIRNA Assays ›
          Gene Symbol:
          Il18
          Gene Name:
          interleukin 18
          Gene Aliases:
          IL-18
          Chromosome Location:
          Chr.8: 55009666 - 55016286 on Build Rnor_6.0
          UniGene ID:
          Rn.11118
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 50608

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Il18
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_019165.1 NP_062038.1 5 500
          GenBank mRNA AJ222813.1 - 5 500
          AY077842.1 - 728
          AY258448.1 - 5 500
          U77776.1 - 5 500
          U77777.1 - 5 443

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_008360, AY362457, AY916131, D49949, DQ316604, and others.

          Panther Classification:

          Molecular Function -

          cytokine interleukin superfamily intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          angiogenesis
          osteoblast differentiation
          response to hypoxia
          inflammatory response
          learning or memory
          response to radiation
          response to cold
          positive regulation of gene expression
          positive regulation of smooth muscle cell migration
          natural killer cell activation
          regulation of cell adhesion
          lung development
          sleep
          lipopolysaccharide-mediated signaling pathway
          positive regulation of chemokine production
          positive regulation of granulocyte macrophage colony-stimulating factor production
          positive regulation of interferon-gamma production
          positive regulation of interleukin-1 beta production
          positive regulation of interleukin-17 production
          positive regulation of interleukin-8 production
          positive regulation of tumor necrosis factor production
          positive regulation of natural killer cell proliferation
          positive regulation of superoxide anion generation
          positive regulation of collagen biosynthetic process
          negative regulation of neutrophil apoptotic process
          T-helper 1 type immune response
          interferon-gamma biosynthetic process
          positive regulation of activated T cell proliferation
          positive regulation of NF-kappaB import into nucleus
          positive regulation of apoptotic process
          positive regulation of vascular permeability
          response to peptide hormone
          positive regulation of neuron apoptotic process
          regulation of circadian sleep/wake cycle, non-REM sleep
          negative regulation of myoblast differentiation
          digestive tract morphogenesis
          positive regulation of smooth muscle cell proliferation
          negative regulation of peptidyl-tyrosine phosphorylation
          detection of mechanical stimulus involved in sensory perception of pain
          positive regulation of NK T cell proliferation
          response to hyperoxia
          cellular response to hydrogen peroxide
          cellular response to lipopolysaccharide
          cellular response to mechanical stimulus
          cellular response to cAMP
          cellular response to interferon-gamma
          cellular response to tumor necrosis factor
          cellular response to parathyroid hormone stimulus
          cellular response to peptide hormone stimulus
          cellular response to organic cyclic compound
          positive regulation of male germ cell proliferation
          positive regulation of blood vessel remodeling
          cytokine activity
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-p5628:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0