Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Hsbp1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136181.1 | NP_001129653.1 | 2, 3 | 287 | ||
GenBank mRNA | AK019003.1 | - | 1, 2 | 141 | ||
AK141935.1 | - | 2, 3 | 288 | |||
BC151118.1 | - | 1, 2 | 134 | 9088031 | ||
BC151120.1 | - | 1, 2 | 134 | 9088033 |