Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Smim20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145433.1 | NP_001138905.1 | 1 | 175 | ||
GenBank mRNA | AK003139.1 | - | 1 | 110 | ||
AK007474.1 | - | 1 | 98 | |||
AK156339.1 | - | 1 | 91 | |||
BC030079.1 | - | 1 | 111 | |||
BY077818.1 | - | 1 | 176 |