Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Prss54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027640.1 | NP_081916.1 | 5 | 979 | ||
GenBank mRNA | AK016501.1 | - | 7 | 1295 | ||
BC144859.1 | - | 6 | 1106 | 9053407 | ||
BC145787.1 | - | 6 | 1106 | 40131090 |