Search
Search

Mfsd12| Gene Symbol: Mfsd12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028657.3 | NP_082933.2 | 2 | 422 | ||
| GenBank mRNA | AK149678.1 | - | 2 | 428 | ||
| AK154386.1 | - | 2 | 424 | |||
| AK154902.1 | - | 2 | 502 | |||
| AK170185.1 | - | 2 | 442 | |||
| AK171084.1 | - | 2 | 424 | |||
| BC118620.1 | - | 2 | 422 | 40107417 | ||
| BC119790.1 | - | 2 | 422 | 40107416 | ||