Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Etnk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029250.2 | NP_083526.2 | 8 | 5673 | ||
GenBank mRNA | AK049691.1 | - | 1 | 3363 | ||
BC023950.1 | - | 2 | 2771 | |||
BC034743.1 | - | 2 | 2701 | |||
BC048829.1 | - | 2 | 1117 |