Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MYL1 SIRNA Assays ›
Gene Symbol:
Myl1
Gene Name:
myosin, light chain 1
Gene Aliases:
MLC3-f, MLCf, Mcl3, Mlc1, Mlc3
Chromosome Location:
Chr.9: 73937820 - 73958480 on Build Rnor_6.0
UniGene ID:
Rn.84920
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
50999
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Myl1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001077656.1
NP_001071124.1
4
486
NM_020104.2
NP_064489.1
4
304
XM_008767294.1
XP_008765516.1
3
434
XM_008767295.1
XP_008765517.1
3
555
GenBank mRNA
CR755322.1
-
4
309
CR756356.1
-
4
304
FQ214580.1
-
4
613
FQ214604.1
-
4
531
FQ214608.1
-
4
491
FQ214631.1
-
4
473
FQ214638.1
-
4
581
FQ214656.1
-
4
322
FQ214675.1
-
4
580
FQ214712.1
-
4
494
FQ214723.1
-
4
494
FQ214727.1
-
4
494
FQ214742.1
-
4
494
FQ214939.1
-
4
491
FQ215002.1
-
4
580
FQ215050.1
-
4
494
FQ215083.1
-
4
491
FQ215086.1
-
4
324
FQ215201.1
-
4
491
FQ215229.1
-
4
585
FQ215325.1
-
4
580
FQ215340.1
-
4
322
FQ215418.1
-
4
580
FQ215435.1
-
4
491
FQ215492.1
-
4
473
FQ215500.1
-
3
1070
FQ215535.1
-
3
555
FQ215557.1
-
4
580
FQ215563.1
-
4
491
FQ215578.1
-
4
580
FQ215748.1
-
3
553
FQ215766.1
-
4
580
FQ215774.1
-
4
491
FQ215820.1
-
4
549
FQ215848.1
-
4
491
FQ215875.1
-
4
491
FQ215911.1
-
4
585
FQ215933.1
-
4
494
FQ216034.1
-
4
494
FQ216060.1
-
4
580
FQ216109.1
-
4
491
FQ216124.1
-
4
585
FQ216181.1
-
4
473
FQ216192.1
-
4
580
FQ216213.1
-
5
488
FQ216218.1
-
4
491
FQ216229.1
-
4
761
FQ216238.1
-
4
580
FQ216308.1
-
4
322
FQ216407.1
-
4
662
FQ216452.1
-
4
491
FQ216455.1
-
4
495
FQ216484.1
-
4
473
FQ216496.1
-
4
494
FQ216525.1
-
4
494
FQ216543.1
-
4
491
FQ216556.1
-
4
580
FQ216567.1
-
4
491
FQ216631.1
-
4
494
FQ216654.1
-
4
578
FQ216704.1
-
4
494
FQ216737.1
-
4
577
FQ216738.1
-
4
322
FQ217074.1
-
4
321
FQ217103.1
-
4
322
FQ217157.1
-
3
297
FQ217192.1
-
4
322
FQ217218.1
-
4
322
FQ217243.1
-
4
321
FQ217303.1
-
4
473
FQ217323.1
-
4
322
FQ217387.1
-
4
322
FQ217394.1
-
4
424
FQ217427.1
-
4
327
FQ217481.1
-
4
322
FQ217524.1
-
4
322
FQ217753.1
-
4
327
FQ217756.1
-
4
322
FQ217889.1
-
4
322
FQ217973.1
-
4
327
FQ217979.1
-
4
322
FQ218029.1
-
4
322
FQ218049.1
-
4
322
FQ223584.1
-
4
580
FQ223632.1
-
4
494
FQ223641.1
-
4
580
FQ223687.1
-
4
322
FQ223755.1
-
4
490
FQ223779.1
-
4
322
FQ223827.1
-
4
494
FQ223880.1
-
4
392
FQ223949.1
-
4
320
FQ223984.1
-
4
322
FQ224047.1
-
4
322
FQ224050.1
-
4
322
FQ224087.1
-
4
322
FQ224155.1
-
4
327
FQ224171.1
-
4
491
FQ224172.1
-
4
322
FQ224212.1
-
4
474
FQ224254.1
-
4
322
FQ224277.1
-
4
494
FQ224321.1
-
4
326
FQ224386.1
-
4
327
FQ224411.1
-
4
322
FQ224420.1
-
4
322
FQ224452.1
-
4
397
FQ224519.1
-
4
491
FQ224609.1
-
5
350
FQ224663.1
-
4
322
FQ224726.1
-
4
424
FQ224796.1
-
4
322
FQ224799.1
-
4
327
FQ224867.1
-
4
580
FQ224927.1
-
4
580
FQ224931.1
-
4
491
FQ224934.1
-
4
489
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
muscle contraction
cardiac muscle contraction
calcium ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0