Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CCR1 SIRNA Assays ›
Gene Symbol:
Ccr1
Gene Name:
chemokine (C-C motif) receptor 1
Gene Aliases:
-
Chromosome Location:
Chr.8: 132996646 - 133002201 on Build Rnor_6.0
UniGene ID:
Rn.34673
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
51182
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Ccr1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_020542.2
NP_065417.1
2
549
GenBank mRNA
BC079207.1
-
2
549
7131790
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_009912, CT010178, BC011092, AK036597, AK031109, and others.
Set Membership:
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
calcium ion transport
cellular calcium ion homeostasis
exocytosis
inflammatory response
immune response
G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
cell-cell signaling
metabolic process
negative regulation of gene expression
cerebellum development
myeloid cell differentiation
negative regulation of bone mineralization
leukocyte chemotaxis
positive regulation of osteoclast differentiation
negative regulation of innate immune response
positive regulation of calcium ion transport
chemokine-mediated signaling pathway
positive regulation of ERK1 and ERK2 cascade
positive regulation of monocyte chemotaxis
phosphatidylinositol phospholipase C activity
chemokine receptor activity
C-C chemokine receptor activity
C-C chemokine binding
chemokine (C-C motif) ligand 7 binding
chemokine (C-C motif) ligand 5 binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline