Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mark2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021699.1 | NP_067731.1 | 4 | 358 | ||
XM_006230879.2 | XP_006230941.1 | 4 | 858 | |||
XM_006230880.2 | XP_006230942.1 | 4 | 1645 | |||
XM_006230881.2 | XP_006230943.1 | 4 | 377 | |||
XM_006230883.2 | XP_006230945.1 | 4 | 857 | |||
XM_006230884.2 | XP_006230946.1 | 4 | 856 | |||
XM_006230885.2 | XP_006230947.1 | 4 | 856 | |||
XM_006230887.2 | XP_006230949.1 | 4 | 855 | |||
XM_008760169.1 | XP_008758391.1 | 4 | 857 | |||
GenBank mRNA | Z83869.1 | - | 4 | 358 |