Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: USP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304284.1 | NP_001291213.1 | 20 | 3112 | ||
NM_004505.3 | NP_004496.2 | 12 | 2581 | |||
XM_011524050.1 | XP_011522352.1 | 16 | 3330 | |||
XM_011524051.2 | XP_011522353.1 | 18 | 2425 | |||
XM_011524052.2 | XP_011522354.1 | 17 | 2539 | |||
XM_011524053.2 | XP_011522355.1 | 18 | 2452 | |||
XM_011524054.2 | XP_011522356.1 | 18 | 2419 | |||
XM_011524055.2 | XP_011522357.1 | 16 | 2346 | |||
XM_011524056.2 | XP_011522358.1 | 13 | 2299 | |||
XM_011524057.2 | XP_011522359.1 | 16 | 2462 | |||
XM_011524058.2 | XP_011522360.1 | 17 | 2528 | |||
XM_011524059.2 | XP_011522361.1 | 17 | 2528 | |||
XM_017025290.1 | XP_016880779.1 | 15 | 3168 | |||
GenBank mRNA | AB449915.1 | - | 11 | 882 | ||
AY143550.1 | - | 12 | 2581 | |||
BX647719.1 | - | 20 | 3113 | |||
X63546.1 | - | 12 | 2578 | |||
X63547.1 | - | 12 | 2578 |