Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FTL1 SIRNA Assays ›
          Gene Symbol:
          Ftl1 Fam65b LOC100360087
          Gene Name:
          ferritin light chain 1
          family with sequence similarity 65, member B
          ferritin light chain 1-like
          Gene Aliases:
          Ftl
          Ab2-162, RGD1306939
          -
          Chromosome Location:
          Chr.1: 101448190 - 101450034 on Build Rnor_6.0
          Chromosome Location:
          Chr.17: 42324594 - 42444935 on Build Rnor_6.0
          Chromosome Location:
          Chr.5: 152325856 - 152326797 on Build Rnor_6.0
          UniGene ID:
          Rn.1905
          Rn.202381
          -
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID 51954

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 286.00
          Your Price
          Online offer: 277.65
          277.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ftl1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_022500.4 NP_071945.3 4 627
          GenBank mRNA BC061525.1 - 4 631 6921603
          BC086583.1 - 4 608 7307718
          BC088756.1 - 4 608 7307747
          FQ209407.1 - 4 627
          FQ209694.1 - 4 627
          FQ209816.1 - 4 627
          FQ218577.1 - 4 627
          FQ219483.1 - 4 627
          FQ219685.1 - 4 627
          FQ219809.1 - 4 627
          FQ219930.1 - 4 626
          FQ220095.1 - 4 627
          FQ220123.1 - 4 627
          FQ220243.1 - 4 624
          FQ220651.1 - 4 627
          FQ220814.1 - 4 627
          FQ220884.1 - 4 627
          FQ221898.1 - 4 627
          FQ222411.1 - 3 531
          FQ226019.1 - 4 627
          FQ226254.1 - 4 583
          FQ226458.1 - 4 626
          FQ226986.1 - 4 623
          FQ227401.1 - 4 627
          FQ227800.1 - 4 627
          FQ227843.1 - 4 629
          FQ228050.1 - 4 627
          FQ228585.1 - 4 627
          FQ229176.1 - 4 627
          FQ229284.1 - 4 627
          FQ229370.1 - 4 627
          FQ229673.1 - 4 625
          FQ229757.1 - 4 627
          FQ230098.1 - 4 625
          FQ230445.1 - 4 627
          FQ230659.1 - 4 623
          FQ231322.1 - 4 627
          FQ231373.1 - 4 627
          FQ231415.1 - 3 483
          FQ231464.1 - 4 627
          FQ231615.1 - 4 627
          FQ231781.1 - 4 623
          FQ232031.1 - 4 627
          FQ232167.1 - 4 627
          FQ232317.1 - 4 583
          FQ232517.1 - 4 627
          FQ233772.1 - 4 628
          FQ233996.1 - 4 627
          FQ234074.1 - 4 627
          FQ234603.1 - 4 627
          FQ234647.1 - 4 627
          FQ235044.1 - 4 627
          FQ235250.1 - 4 627
          K01930.1 - 4 552
          L01122.1 - 4 580
          Gene Symbol: Fam65b
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001014009.2 NP_001014031.2 27 3803
          GenBank mRNA AY325194.1 - 27 3803
          Gene Symbol: LOC100360087
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_002729553.4 XP_002729599.1 1 642

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: S89400.

          Panther Classification:

          Molecular Function -

          storage protein non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          iron ion transport
          cellular iron ion homeostasis
          response to lead ion
          sensory perception of sound
          molecular_function
          iron ion binding
          ferric iron binding
          identical protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0