Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Bbox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022629.1 | NP_072151.1 | 1, 2 | 227 | ||
GenBank mRNA | AF160958.1 | - | 1, 2 | 227 | ||
FQ210746.1 | - | 319 |