Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other B3GAT2 SIRNA Assays ›
Gene Symbol:
B3gat2
Gene Name:
beta-1,3-glucuronyltransferase 2
Gene Aliases:
-
Chromosome Location:
Chr.9: 29984077 - 30068649 on Build Rnor_6.0
UniGene ID:
Rn.42869
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
52114
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: B3gat2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_022609.2
NP_072131.1
3
1458
GenBank mRNA
AB010441.1
-
3
1458
AF106624.1
-
3
1453
Back To Top
More Information
Panther Classification:
Molecular Function -
glycosyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
protein glycosylation
metabolic process
carbohydrate biosynthetic process
galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline