Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022847.1 | NP_074038.1 | 3, 4 | 1767 | ||
GenBank mRNA | HM037361.1 | - | 2319 | |||
HM037362.1 | - | 2319 | ||||
L16922.1 | - | 3, 4 | 1767 |