Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Creb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031017.1 | NP_112279.1 | 8 | 908 | ||
NM_134443.1 | NP_604392.1 | 6 | 827 | |||
XM_006245065.2 | XP_006245127.1 | 8 | 914 | |||
XM_006245066.2 | XP_006245128.1 | 7 | 1079 | |||
XM_006245067.2 | XP_006245129.1 | 7 | 819 | |||
XM_006245068.2 | XP_006245130.1 | 6 | 766 | |||
XM_006245069.2 | XP_006245131.1 | 6 | 1416 | |||
XM_006245070.2 | XP_006245132.1 | 3 | 286 | |||
XM_008767138.1 | XP_008765360.1 | 5 | 3270 | |||
XM_008767139.1 | XP_008765361.1 | 2 | 1140 | |||
GenBank mRNA | BC097311.1 | - | 9 | 1172 | ||
FQ211925.1 | - | 8 | 973 | |||
X14788.1 | - | 8 | 908 | |||
X60002.1 | - | 6 | 827 |
Gene Symbol: LOC100363500 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772050.1 | XP_008770272.1 | 1 | 862 |