Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Smcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031536.2 | NP_113724.1 | 2 | 582 | ||
GenBank mRNA | BC078795.1 | - | 2 | 493 | 7134574 | |
U48702.1 | - | 2 | 588 | |||
X87883.1 | - | 2 | 574 |