Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CCL2 SIRNA Assays ›
Gene Symbol:
Ccl2
Gene Name:
chemokine (C-C motif) ligand 2
Gene Aliases:
MCP-1, Scya2, Sigje
Chromosome Location:
Chr.10: 69412065 - 69413863 on Build Rnor_6.0
UniGene ID:
Rn.4772
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
53807
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Ccl2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_031530.1
NP_113718.1
3
412
GenBank mRNA
AF058786.1
-
3
412
M57441.1
-
3
376
Back To Top
More Information
Panther Classification:
Molecular Function -
cytokine
intercellular signal molecule
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
response to hypoxia
positive regulation of leukocyte mediated cytotoxicity
positive regulation of endothelial cell proliferation
leukocyte migration involved in inflammatory response
monocyte chemotaxis
positive regulation of leukocyte migration
positive regulation of cellular extravasation
cellular calcium ion homeostasis
inflammatory response
cytoskeleton organization
transforming growth factor beta receptor signaling pathway
aging
glial cell migration
regulation of cell shape
response to heat
response to wounding
response to mechanical stimulus
response to bacterium
response to gamma radiation
regulation of vascular endothelial growth factor production
positive regulation of macrophage chemotaxis
response to activity
negative regulation of angiogenesis
cytokine-mediated signaling pathway
positive regulation of cell-cell adhesion
neutrophil chemotaxis
organ regeneration
lipopolysaccharide-mediated signaling pathway
response to lipopolysaccharide
response to progesterone
positive regulation of tumor necrosis factor production
cellular response to insulin stimulus
positive regulation of collagen biosynthetic process
response to vitamin B3
negative regulation of glial cell apoptotic process
helper T cell extravasation
cellular response to drug
cellular response to macrophage colony-stimulating factor stimulus
cellular response to platelet-derived growth factor stimulus
chemokinesis
response to drug
response to amino acid
protein kinase B signaling
negative regulation of neuron apoptotic process
astrocyte cell migration
cellular response to fibroblast growth factor stimulus
response to ethanol
positive regulation of cell adhesion
response to antibiotic
vascular endothelial growth factor receptor signaling pathway
macrophage chemotaxis
lymphocyte chemotaxis
positive regulation of synaptic transmission
positive regulation of T cell activation
response to glucocorticoid
positive regulation of nitric-oxide synthase biosynthetic process
maternal process involved in female pregnancy
maternal process involved in parturition
chemokine-mediated signaling pathway
cellular response to lipopolysaccharide
cellular response to retinoic acid
cellular response to ATP
cellular response to interferon-gamma
cellular response to interleukin-1
cellular response to interleukin-6
cellular response to tumor necrosis factor
cellular response to lipoprotein particle stimulus
cellular response to high density lipoprotein particle stimulus
cellular response to organic cyclic compound
cellular response to dexamethasone stimulus
positive regulation of monocyte chemotaxis
positive regulation of immune complex clearance by monocytes and macrophages
positive regulation of calcium ion import
positive regulation of protein targeting to membrane
positive regulation of apoptotic cell clearance
negative regulation of natural killer cell chemotaxis
G-protein coupled receptor binding
chemokine activity
heparin binding
CCR2 chemokine receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0