Search
Search

CDKN2B| Gene Symbol: CDKN2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004936.3 | NP_004927.2 | 2 | 1072 | ||
| NM_078487.2 | NP_511042.1 | 2 | 1195 | |||
| GenBank mRNA | AF488409.1 | - | 1 | 580 | ||
| AF488410.1 | - | 1 | 353 | |||
| BC014469.1 | - | 2 | 1040 | 4871014 | ||