Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031853.4 | NP_114054.1 | 3, 4 | 315 | ||
GenBank mRNA | AY383701.1 | - | 495 | |||
BC084717.1 | - | 2, 3 | 202 | 7310272 | ||
FQ209835.1 | - | 3, 4 | 263 | |||
FQ209938.1 | - | 3, 4 | 258 | |||
FQ210140.1 | - | 3, 4 | 263 | |||
FQ210165.1 | - | 3, 4 | 258 | |||
FQ210336.1 | - | 3, 4 | 260 | |||
FQ210460.1 | - | 3, 4 | 263 | |||
FQ210781.1 | - | 3, 4 | 290 | |||
FQ210997.1 | - | 3, 4 | 260 | |||
FQ211081.1 | - | 3, 4 | 263 | |||
FQ211192.1 | - | 3, 4 | 263 | |||
FQ211339.1 | - | 3, 4 | 264 | |||
FQ211377.1 | - | 2, 3 | 184 | |||
FQ213527.1 | - | 3, 4 | 264 | |||
FQ214572.1 | - | 3, 4 | 263 | |||
FQ217714.1 | - | 3, 4 | 264 | |||
FQ218148.1 | - | 3, 4 | 258 | |||
FQ218164.1 | - | 3, 4 | 282 | |||
FQ218239.1 | - | 3, 4 | 263 | |||
FQ218300.1 | - | 3, 4 | 263 | |||
FQ218680.1 | - | 3, 4 | 263 | |||
FQ219136.1 | - | 3, 4 | 263 | |||
FQ219739.1 | - | 3, 4 | 263 | |||
FQ221153.1 | - | 3, 4 | 258 | |||
FQ221871.1 | - | 3, 4 | 264 | |||
FQ222052.1 | - | 3, 4 | 263 | |||
FQ222378.1 | - | 3, 4 | 265 | |||
FQ222416.1 | - | 3, 4 | 258 | |||
FQ222549.1 | - | 3, 4 | 253 | |||
FQ222564.1 | - | 3, 4 | 264 | |||
FQ222579.1 | - | 3, 4 | 294 | |||
FQ223167.1 | - | 3, 4 | 258 | |||
FQ223413.1 | - | 3, 4 | 263 | |||
FQ223492.1 | - | 3, 4 | 264 | |||
FQ223501.1 | - | 3, 4 | 263 | |||
FQ223602.1 | - | 3, 4 | 221 | |||
FQ224361.1 | - | 3, 4 | 263 | |||
FQ224797.1 | - | 3, 4 | 264 | |||
FQ229114.1 | - | 3, 4 | 258 | |||
M14201.1 | - | 3, 4 | 315 | |||
M20268.1 | - | 3, 4 | 204 |