Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pawr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033485.2 | NP_277020.1 | 1 | 489 | ||
XM_006241317.2 | XP_006241379.1 | 2 | 702 | |||
GenBank mRNA | U05989.1 | - | 1 | 489 |