Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Dbi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031853.4 | NP_114054.1 | 4 | 356 | ||
GenBank mRNA | BC084717.1 | - | 3 | 243 | 7310272 | |
FQ209769.1 | - | 4 | 282 | |||
FQ209835.1 | - | 4 | 304 | |||
FQ209938.1 | - | 4 | 299 | |||
FQ210140.1 | - | 4 | 304 | |||
FQ210165.1 | - | 4 | 299 | |||
FQ210336.1 | - | 4 | 301 | |||
FQ210460.1 | - | 4 | 304 | |||
FQ210781.1 | - | 4 | 331 | |||
FQ210997.1 | - | 4 | 301 | |||
FQ211081.1 | - | 4 | 304 | |||
FQ211136.1 | - | 4 | 304 | |||
FQ211192.1 | - | 4 | 304 | |||
FQ211339.1 | - | 4 | 305 | |||
FQ211377.1 | - | 3 | 225 | |||
FQ213527.1 | - | 4 | 305 | |||
FQ214572.1 | - | 4 | 304 | |||
FQ217714.1 | - | 4 | 305 | |||
FQ218148.1 | - | 4 | 299 | |||
FQ218164.1 | - | 4 | 323 | |||
FQ218239.1 | - | 4 | 304 | |||
FQ218300.1 | - | 4 | 304 | |||
FQ219136.1 | - | 4 | 304 | |||
FQ219739.1 | - | 4 | 304 | |||
FQ221153.1 | - | 4 | 299 | |||
FQ221871.1 | - | 4 | 305 | |||
FQ222052.1 | - | 4 | 304 | |||
FQ222378.1 | - | 4 | 306 | |||
FQ222416.1 | - | 4 | 299 | |||
FQ222549.1 | - | 4 | 294 | |||
FQ222564.1 | - | 4 | 305 | |||
FQ223167.1 | - | 4 | 299 | |||
FQ223413.1 | - | 4 | 304 | |||
FQ223492.1 | - | 4 | 305 | |||
FQ223501.1 | - | 4 | 304 | |||
FQ223602.1 | - | 4 | 262 | |||
FQ224361.1 | - | 4 | 304 | |||
FQ224797.1 | - | 4 | 305 | |||
FQ229114.1 | - | 4 | 299 | |||
M14201.1 | - | 4 | 356 | |||
M20268.1 | - | 4 | 245 |