Search
Search

Bnip3| Gene Symbol: Bnip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053420.3 | NP_445872.1 | 4, 5 | 498 | ||
| GenBank mRNA | AF243515.1 | - | 4, 5 | 462 | ||
| AY095482.1 | - | 4, 5 | 462 | |||
| BC070958.1 | - | 4, 5 | 456 | 7105707 | ||
| BC081690.1 | - | 4, 5 | 436 | 7190121 | ||
| CO403082.1 | - | 4, 5 | 534 | |||
| FQ213448.1 | - | 3, 4 | 259 | |||
| FQ215108.1 | - | 4, 5 | 468 | |||
| FQ215127.1 | - | 4, 5 | 468 | |||
| FQ215203.1 | - | 4, 5 | 489 | |||
| FQ216745.1 | - | 4, 5 | 489 | |||
| FQ216768.1 | - | 1, 2 | 75 | |||
| FQ216807.1 | - | 4, 5 | 489 | |||
| FQ220853.1 | - | 4, 5 | 467 | |||
| FQ221849.1 | - | 4, 5 | 489 | |||
| KC660101.1 | - | 3, 4 | 295 | |||