Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPS6KA3 SIRNA Assays ›
          Gene Symbol:
          RPS6KA3
          Gene Name:
          ribosomal protein S6 kinase A3
          Gene Aliases:
          CLS, HU-3, ISPK-1, MAPKAPK1B, MRX19, RSK, RSK2, S6K-alpha3, p90-RSK2, pp90RSK2
          Chromosome Location:
          Chr.X: 20149911 - 20267514 on Build GRCh38
          UniGene ID:
          Hs.445387
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 553
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # AM51331
          Price 6,301.00
          Your Price
          Online offer:
          6301.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RPS6KA3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_004586.2 NP_004577.1 3 183
          XM_005274573.2 XP_005274630.1 3 187
          XM_005274577.2 XP_005274634.1 2 123
          XM_006724507.3 XP_006724570.1 5 289
          XM_011545555.1 XP_011543857.1 3 187
          XM_011545556.1 XP_011543858.1 3 187
          XM_011545557.2 XP_011543859.1 5 305
          XM_011545558.2 XP_011543860.1 5 369
          XM_011545559.1 XP_011543861.1 5 366
          XM_011545560.1 XP_011543862.1 4 221
          XM_011545561.1 XP_011543863.1 3 147
          XM_011545562.1 XP_011543864.1 2 123
          XM_011545563.2 XP_011543865.1 5 303
          XM_017029713.1 XP_016885202.1 5 291
          XM_017029714.1 XP_016885203.1 5 367
          XM_017029715.1 XP_016885204.1 5 366
          XM_017029716.1 XP_016885205.1 5 441
          XM_017029717.1 XP_016885206.1 3 147
          XM_017029718.1 XP_016885207.1 2 123
          XM_017029719.1 XP_016885208.1 3 147
          GenBank mRNA AB102662.1 - 3 142
          AB208933.1 - 3 385
          AK294373.1 - 5 278
          AK296775.1 - 2 119
          AK313932.1 - 3 187
          AK316482.1 - 4 582
          BC096301.1 - 3 183 40007452
          BC096302.3 - 3 183 40007453
          BC096303.1 - 3 183 40007456
          L07599.1 - 3 180
          U08316.1 - 3 183

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001192004, XM_008773193, XM_008773189, XM_008773190, XM_008773188, and others.
          This siRNA also targets the following transcripts in Mouse: NM_148945, AY083469, BC150156, BC150478, AK079102, and others.

          Matching Taqman Gene Expression Assay:


          Hs00177936_m1

          Panther Classification:

          Molecular Function -

          protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal system development
          toll-like receptor signaling pathway
          protein phosphorylation
          apoptotic process
          cell cycle
          signal transduction
          central nervous system development
          positive regulation of cell growth
          response to lipopolysaccharide
          intracellular signal transduction
          negative regulation of apoptotic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          regulation of translation in response to stress
          regulation of DNA-templated transcription in response to stress
          positive regulation of cell differentiation
          positive regulation of transcription from RNA polymerase II promoter
          magnesium ion binding
          protein kinase activity
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          kinase activity
          protein kinase binding
          cysteine-type endopeptidase inhibitor activity involved in apoptotic process

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0