Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RPS6KA3 SIRNA Assays ›
Gene Symbol:
RPS6KA3
Gene Name:
ribosomal protein S6 kinase A3
Gene Aliases:
CLS, HU-3, ISPK-1, MAPKAPK1B, MRX19, RSK, RSK2, S6K-alpha3, p90-RSK2, pp90RSK2
Chromosome Location:
Chr.X: 20149911 - 20267514 on Build GRCh38
UniGene ID:
Hs.445387
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
554
Validated
Purification
In-Vivo Ready
Standard
HPLC
Size
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
Availability
Inventoried
Catalog #
AM51331
Price
6,301.00
Your Price
Online offer:
6301.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RPS6KA3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_004586.2
NP_004577.1
18, 19
1772
XM_005274573.2
XP_005274630.1
18, 19
1773
XM_005274577.2
XP_005274634.1
17, 18
1709
XM_006724507.3
XP_006724570.1
20, 21
1875
XM_011545555.1
XP_011543857.1
18, 19
1794
XM_011545556.1
XP_011543858.1
18, 19
1791
XM_011545557.2
XP_011543859.1
20, 21
1912
XM_011545558.2
XP_011543860.1
20, 21
1976
XM_011545559.1
XP_011543861.1
20, 21
1973
XM_011545560.1
XP_011543862.1
19, 20
1828
XM_011545561.1
XP_011543863.1
18, 19
1754
XM_011545562.1
XP_011543864.1
17, 18
1730
XM_011545563.2
XP_011543865.1
20, 21
1892
XM_017029713.1
XP_016885202.1
20, 21
1880
XM_017029714.1
XP_016885203.1
20, 21
1956
XM_017029715.1
XP_016885204.1
20, 21
1955
XM_017029716.1
XP_016885205.1
20, 21
2030
XM_017029717.1
XP_016885206.1
18, 19
1736
XM_017029718.1
XP_016885207.1
17, 18
1712
XM_017029719.1
XP_016885208.1
18, 19
1733
GenBank mRNA
AB102662.1
-
18, 19
1731
AB208933.1
-
18, 19
1971
AK294373.1
-
20, 21
1864
AK296775.1
-
17, 18
1705
AK313932.1
-
18, 19
1776
AK316482.1
-
19, 20
2171
BC096301.1
-
18, 19
1772
40007452
BC096302.3
-
18, 19
1772
40007453
BC096303.1
-
18, 19
1772
40007456
U08316.1
-
18, 19
1772
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00177936_m1
Panther Classification:
Molecular Function -
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
skeletal system development
toll-like receptor signaling pathway
protein phosphorylation
apoptotic process
cell cycle
signal transduction
central nervous system development
positive regulation of cell growth
response to lipopolysaccharide
intracellular signal transduction
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
regulation of translation in response to stress
regulation of DNA-templated transcription in response to stress
positive regulation of cell differentiation
positive regulation of transcription from RNA polymerase II promoter
magnesium ion binding
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
kinase activity
protein kinase binding
cysteine-type endopeptidase inhibitor activity involved in apoptotic process
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0