Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPS6KA3 SIRNA Assays ›
          Gene Symbol:
          RPS6KA3
          Gene Name:
          ribosomal protein S6 kinase A3
          Gene Aliases:
          CLS, HU-3, ISPK-1, MAPKAPK1B, MRX19, RSK, RSK2, S6K-alpha3, p90-RSK2, pp90RSK2
          Chromosome Location:
          Chr.X: 20149911 - 20267514 on Build GRCh38
          UniGene ID:
          Hs.445387
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 555
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # AM51331
          Price 6,301.00
          Your Price
          Online offer:
          6301.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RPS6KA3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_004586.2 NP_004577.1 2 113
          XM_005274573.2 XP_005274630.1 2 117
          XM_006724507.3 XP_006724570.1 4 219
          XM_011545555.1 XP_011543857.1 2 117
          XM_011545556.1 XP_011543858.1 2 117
          XM_011545557.2 XP_011543859.1 4 235
          XM_011545558.2 XP_011543860.1 4 299
          XM_011545559.1 XP_011543861.1 4 296
          XM_011545560.1 XP_011543862.1 3 151
          XM_011545561.1 XP_011543863.1 2 77
          XM_011545563.2 XP_011543865.1 4 233
          XM_017029713.1 XP_016885202.1 4 221
          XM_017029714.1 XP_016885203.1 4 297
          XM_017029715.1 XP_016885204.1 4 296
          XM_017029716.1 XP_016885205.1 4 371
          XM_017029717.1 XP_016885206.1 2 77
          XM_017029719.1 XP_016885208.1 2 77
          GenBank mRNA AB102662.1 - 2 72
          AB208933.1 - 2 315
          AK294373.1 - 4 208
          AK313932.1 - 2 117
          AK316482.1 - 3 512
          BC096301.1 - 2 113 40007452
          BC096302.3 - 2 113 40007453
          BC096303.1 - 2 113 40007456
          L07599.1 - 2 110
          U08316.1 - 2 113

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_148945, AY083469, BC150478, CJ123041, BC150156, and others.

          Matching Taqman Gene Expression Assay:


          Hs00177936_m1

          Panther Classification:

          Molecular Function -

          protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal system development
          toll-like receptor signaling pathway
          protein phosphorylation
          apoptotic process
          cell cycle
          signal transduction
          central nervous system development
          positive regulation of cell growth
          response to lipopolysaccharide
          intracellular signal transduction
          negative regulation of apoptotic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          regulation of translation in response to stress
          regulation of DNA-templated transcription in response to stress
          positive regulation of cell differentiation
          positive regulation of transcription from RNA polymerase II promoter
          magnesium ion binding
          protein kinase activity
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          kinase activity
          protein kinase binding
          cysteine-type endopeptidase inhibitor activity involved in apoptotic process

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0