Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
CDMO and Clinical Trial Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZHX3 SIRNA Assays ›
Gene Symbol:
ZHX3
Gene Name:
zinc fingers and homeoboxes 3
Gene Aliases:
TIX1
Chromosome Location:
Chr.20: 41178449 - 41317606 on Build GRCh38
UniGene ID:
Hs.380133
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
5621
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZHX3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_015035.3
NP_055850.1
3
914
XM_005260341.3
XP_005260398.1
3
941
XM_005260343.4
XP_005260400.1
3
941
XM_006723748.3
XP_006723811.1
4
1002
XM_006723749.3
XP_006723812.1
2
825
XM_011528701.2
XP_011527003.1
3
878
XM_011528702.2
XP_011527004.1
6
1194
XM_011528703.2
XP_011527005.1
5
1146
XM_011528704.2
XP_011527006.1
3
941
XM_011528705.2
XP_011527007.1
3
957
XM_011528706.2
XP_011527008.1
3
877
XM_011528707.2
XP_011527009.1
3
930
XM_011528708.2
XP_011527010.1
3
800
XM_011528709.2
XP_011527011.1
3
802
XM_011528711.2
XP_011527013.1
3
949
XM_011528713.2
XP_011527015.1
4
1039
XM_011528714.2
XP_011527016.1
4
978
XM_011528717.2
XP_011527019.1
4
2653
XM_011528718.2
XP_011527020.1
4
952
XM_011528720.1
XP_011527022.1
3
949
XM_017027736.1
XP_016883225.1
3
855
XM_017027737.1
XP_016883226.1
3
957
XM_017027738.1
XP_016883227.1
3
1080
GenBank mRNA
AB007855.3
-
3
885
AB081948.1
-
3
896
AK097523.1
-
4
952
AK122905.1
-
3
895
AK292415.1
-
3
912
BC068569.1
-
4
999
5295555
Back To Top
More Information
Panther Classification:
Molecular Function -
homeodomain transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
cell differentiation
positive regulation of osteoblast differentiation
negative regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription corepressor activity
protein binding
protein homodimerization activity
metal ion binding
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-68wbn:80/10.239.160.160:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0