Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Hadha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130826.2 | NP_570839.2 | 4, 5 | 325 | ||
GenBank mRNA | BC091697.1 | - | 4, 5 | 325 | 7301707 | |
D16478.1 | - | 4, 5 | 323 | |||
X98225.1 | - | 2, 3 | 211 |