Search
Search

Slc25a21| Gene Symbol: Slc25a21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133614.2 | NP_598298.1 | 1, 2 | 330 | ||
| XM_006240114.2 | XP_006240176.1 | 1, 2 | 1656 | |||
| GenBank mRNA | AJ289714.1 | - | 1, 2 | 174 | ||
| BC089099.1 | - | 1, 2 | 330 | 7306738 | ||