Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Nbn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138873.2 | NP_620228.1 | 10 | 1241 | ||
XM_006237916.1 | XP_006237978.1 | 10 | 1284 | |||
GenBank mRNA | AF218575.1 | - | 10 | 1245 | ||
BC085700.1 | - | 10 | 1209 | 7130978 |