Search
Search

Gng8| Gene Symbol: Gng8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139185.1 | NP_631924.1 | 4 | 363 | ||
| XM_006228384.2 | XP_006228446.1 | 3 | 717 | |||
| XM_006228385.2 | XP_006228447.1 | 3 | 161 | |||
| GenBank mRNA | L35921.1 | - | 4 | 363 | ||