Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MBD1 SIRNA Assays ›
Gene Symbol:
MBD1
Gene Name:
methyl-CpG binding domain protein 1
Gene Aliases:
CXXC3, PCM1, RFT
Chromosome Location:
Chr.18: 50266882 - 50281774 on Build GRCh38
UniGene ID:
Hs.405610
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
5819
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MBD1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001204136.1
NP_001191065.1
16
2247
NM_001204142.1
NP_001191071.1
14
2314
NM_001204143.1
NP_001191072.1
14
1715
NM_001204151.2
NP_001191080.1
15
2209
NM_001323942.1
NP_001310871.1
17
2322
NM_001323947.1
NP_001310876.1
17
2277
NM_001323949.1
NP_001310878.1
18
2374
NM_001323950.1
NP_001310879.1
17
2271
NM_001323952.1
NP_001310881.1
16
2076
NM_001323954.1
NP_001310883.1
14
2010
NM_002384.2
NP_002375.1
15
2013
NM_015844.2
NP_056669.2
16
2151
NM_015845.3
NP_056670.2
14
2040
NM_015846.3
NP_056671.2
17
2319
NM_015847.3
NP_056723.2
16
2172
XM_005258271.2
XP_005258328.1
16
3716
XM_006722456.2
XP_006722519.1
17
2062
XM_011525991.1
XP_011524293.1
16
1931
XM_011525993.2
XP_011524295.1
18
2141
XM_011525994.2
XP_011524296.1
18
2134
XM_011525999.1
XP_011524301.1
15
1856
XM_011526001.1
XP_011524303.1
15
1826
XM_011526002.1
XP_011524304.1
16
3653
XM_011526003.2
XP_011524305.1
17
2003
XM_011526006.1
XP_011524308.1
14
1681
XM_011526007.1
XP_011524309.1
15
3578
XM_017025751.1
XP_016881240.1
17
2017
XM_017025752.1
XP_016881241.1
16
1949
XM_017025753.1
XP_016881242.1
16
1987
XM_017025756.1
XP_016881245.1
16
1924
XM_017025757.1
XP_016881246.1
17
3746
XM_017025758.1
XP_016881247.1
18
2096
XM_017025759.1
XP_016881248.1
18
2089
XM_017025760.1
XP_016881249.1
17
2059
XM_017025762.1
XP_016881251.1
17
2014
XM_017025763.1
XP_016881252.1
16
3668
XM_017025764.1
XP_016881253.1
17
1996
XM_017025765.1
XP_016881254.1
15
1819
XM_017025766.1
XP_016881255.1
17
1973
XM_017025767.1
XP_016881256.1
17
1966
XM_017025769.1
XP_016881258.1
16
1928
XM_017025770.1
XP_016881259.1
15
3548
XM_017025771.1
XP_016881260.1
16
1891
XM_017025773.1
XP_016881262.1
14
3479
XM_017025774.1
XP_016881263.1
15
1829
XM_017025776.1
XP_016881265.1
14
3410
GenBank mRNA
AF078831.1
-
13
1603
AJ564845.1
-
14
2016
AK225889.1
-
16
2157
AK295401.1
-
18
2126
AK300759.1
-
14
1715
BC033242.1
-
16
1877
5432943
Y10746.1
-
16
1874
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
methylation-dependent chromatin silencing
transcription from RNA polymerase II promoter
heart development
aging
response to fluoxetine
neuron differentiation
response to nutrient levels
response to estradiol
response to cocaine
regulation of DNA methylation
negative regulation of transcription, DNA-templated
negative regulation of astrocyte differentiation
DNA binding
chromatin binding
protein binding
zinc ion binding
methyl-CpG binding
double-stranded methylated DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-xjq9k:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0